gRNA Design Details
- Target DNA Sequence
- CGGGATCTGGATGTCATCAC
- PAM Sequence
- TGG
- Target locus
-
Chr.17: 89200550 - 89200572 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
88.41/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Fshr
- Entrez Gene ID
-
14309
- Gene Name
- follicle stimulating hormone receptor
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 88984952 - 89200675 on GRCm38
- UniGene ID
- Mm.57155
- Species
-
Mouse