gRNA Design Details
- Target DNA Sequence
- CCCTGAATGATCACCTGGTG
- PAM Sequence
- GGG
- Target locus
-
Chr.3: 89890259 - 89890281 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.27/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Il6ra
- Entrez Gene ID
-
16194
- Gene Name
- interleukin 6 receptor, alpha
- Gene Aliases
-
CD126, IL-6R, Il6r
- Chromosome Location
-
Chr.3: 89868829 - 89913492 on GRCm38
- UniGene ID
- Mm.2856
- Species
-
Mouse