gRNA Design Details
- Target DNA Sequence
- CATTGGATTTCTCGCGCTCG
- PAM Sequence
- CGG
- Application
- Gene Knockout
Score:
92.09/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Fgf13
- Entrez Gene ID
-
14168
- Gene Name
- fibroblast growth factor 13
- Gene Aliases
-
Fhf2
- Chromosome Location
-
Chr.X: 59062139 - 59585572 on GRCm38
- UniGene ID
- Mm.7995
- Species
-
Mouse