gRNA Design Details
- Target DNA Sequence
- CCCTACATTGTGAAGCTCCT
- PAM Sequence
- GGG
- Target locus
-
Chr.5: 172156652 - 172156674 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
88.06/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- STK10
- Entrez Gene ID
-
6793
- Gene Name
- serine/threonine kinase 10
- Gene Aliases
-
LOK, PRO2729
- Chromosome Location
-
Chr.5: 172042070 - 172188342 on GRCh38
- UniGene ID
- Hs.744005
- Species
-
Human