gRNA Design Details

Target DNA Sequence
CTACGTGTTGTTGCCATCCA
PAM Sequence
GGG
Target locus
Chr.13: 101868780 - 101868802 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 92.12/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
FGF14
Entrez Gene ID
2259
Gene Name
fibroblast growth factor 14
Gene Aliases
FGF-14, FHF-4, FHF4, SCA27
Chromosome Location
Chr.13: 101720855 - 102402428 on GRCh38
UniGene ID
Hs.508616
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001321931.1 NP_001308860.1 4
NM_001321932.1 NP_001308861.1 4
NM_001321933.1 NP_001308862.1 4
NM_001321934.1 NP_001308863.1 3
NM_001321935.1 NP_001308864.1 3
NM_001321936.1 NP_001308865.1 4
NM_001321937.1 NP_001308866.1 3
NM_001321938.1 NP_001308867.1 5
NM_001321939.1 NP_001308868.1 2
NM_001321940.1 NP_001308869.1 4
NM_001321941.1 NP_001308870.1 4
NM_001321942.1 NP_001308871.1 4
NM_001321943.1 NP_001308872.1 4
NM_001321944.1 NP_001308873.1 4
NM_001321945.1 NP_001308874.1 4
NM_001321946.1 NP_001308875.1 5
NM_001321947.1 NP_001308876.1 4
NM_001321948.1 NP_001308877.1 4
NM_001321949.1 NP_001308878.1 4
NM_004115.3 NP_004106.1 3
NM_175929.2 NP_787125.1 3