gRNA Design Details
- Target DNA Sequence
- ACGGAGACCACTCTTCACGA
- PAM Sequence
- TGG
- Target locus
-
Chr.1: 156864730 - 156864752 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
94/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- NTRK1
- Entrez Gene ID
-
4914
- Gene Name
- neurotrophic receptor tyrosine kinase 1
- Gene Aliases
-
MTC, TRK, TRK1, TRKA, Trk-A, p140-TrkA
- Chromosome Location
-
Chr.1: 156815750 - 156881850 on GRCh38
- UniGene ID
- Hs.406293
- Species
-
Human