gRNA Design Details
- Target DNA Sequence
- ATCAGCTACGAGTGCTGTCC
- PAM Sequence
- TGG
- Target locus
-
Chr.5: 136044062 - 136044084 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
90.85/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TGFBI
- Entrez Gene ID
-
7045
- Gene Name
- transforming growth factor beta induced
- Gene Aliases
-
BIGH3, CDB1, CDG2, CDGG1, CSD, CSD1, CSD2, CSD3, EBMD, LCD1
- Chromosome Location
-
Chr.5: 136028895 - 136063818 on GRCh38
- UniGene ID
- Hs.369397
- Species
-
Human