gRNA Design Details
- Target DNA Sequence
- TAATCTGGGCTGTTTTCGGA
- PAM Sequence
- AGG
- Target locus
-
Chr.2: 55917950 - 55917972 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
88.99/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- EFEMP1
- Entrez Gene ID
-
2202
- Gene Name
- EGF containing fibulin like extracellular matrix protein 1
- Gene Aliases
-
DHRD, DRAD, FBLN3, FBNL, FIBL-3, MLVT, MTLV, S1-5
- Chromosome Location
-
Chr.2: 55865962 - 55924163 on GRCh38
- UniGene ID
- Hs.732348
- Species
-
Human