gRNA Design Details
- Target DNA Sequence
- GGTGTTCTCTCAACATCTCC
- PAM Sequence
- CGG
- Target locus
-
Chr.2: 165657963 - 165657985 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.22/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CSRNP3
- Entrez Gene ID
-
80034
- Gene Name
- cysteine and serine rich nuclear protein 3
- Gene Aliases
-
FAM130A2, PPP1R73, TAIP-2, TAIP2
- Chromosome Location
-
Chr.2: 165469647 - 165689407 on GRCh38
- UniGene ID
- Hs.470479
- Species
-
Human