gRNA Design Details

Target DNA Sequence
GTCTGCAGTATTCACTTTTT
PAM Sequence
TGG
Target locus
Chr.7: 81762757 - 81762779 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 81.88/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
HGF
Entrez Gene ID
3082
Gene Name
hepatocyte growth factor
Gene Aliases
DFNB39, F-TCF, HGFB, HPTA, SF
Chromosome Location
Chr.7: 81699006 - 81770438 on GRCh38
UniGene ID
Hs.396530
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_000601.5 NP_000592.3 2
NM_001010931.2 NP_001010931.1 2
NM_001010932.2 NP_001010932.1 2
NM_001010933.2 NP_001010933.1 2
NM_001010934.2 NP_001010934.1 2
XM_006715956.2 XP_006716019.1 3
XM_011516115.2 XP_011514417.1 3
XM_017012097.1 XP_016867586.1 3
XM_017012098.1 XP_016867587.1 3