gRNA Design Details
- Target DNA Sequence
- GGCCGTAGTACCATTCGTTG
- PAM Sequence
- TGG
- Target locus
-
Chr.2: 75198631 - 75198653 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
95.09/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TACR1
- Entrez Gene ID
-
6869
- Gene Name
- tachykinin receptor 1
- Gene Aliases
-
NK1R, NKIR, SPR, TAC1R
- Chromosome Location
-
Chr.2: 75046463 - 75199519 on GRCh38
- UniGene ID
- Hs.633301
- Species
-
Human