gRNA Design Details
- Target DNA Sequence
- CAATTACTCGCTCACCACAA
- PAM Sequence
- AGG
- Target locus
-
Chr.1: 29160426 - 29160448 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
89.13/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SRSF4
- Entrez Gene ID
-
6429
- Gene Name
- serine and arginine rich splicing factor 4
- Gene Aliases
-
SFRS4, SRP75
- Chromosome Location
-
Chr.1: 29147738 - 29182125 on GRCh38
- UniGene ID
- Hs.469970
- Species
-
Human