gRNA Design Details

Target DNA Sequence
GGACGAACTGGACAGTAACA
PAM Sequence
TGG
Target locus
Chr.19: 48955801 - 48955823 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 89.99/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
BAX
Entrez Gene ID
581
Gene Name
BCL2 associated X, apoptosis regulator
Gene Aliases
BCL2L4
Chromosome Location
Chr.19: 48954825 - 48961798 on GRCh38
UniGene ID
Hs.624291
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001291428.1 NP_001278357.1 3
NM_001291429.1 NP_001278358.1 2
NM_004324.3 NP_004315.1 3
NM_138761.3 NP_620116.1 3
NM_138764.4 NP_620119.2 3
NR_027882.1 3
XM_017027077.1 XP_016882566.1 3