gRNA Design Details

Target DNA Sequence
TTGGATTGTTGGATAACCCT
PAM Sequence
AGG
Target locus
Chr.9: 92080876 - 92080898 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 87.96/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
SPTLC1
Entrez Gene ID
10558
Gene Name
serine palmitoyltransferase long chain base subunit 1
Gene Aliases
HSAN1, HSN1, LBC1, LCB1, SPT1, SPTI
Chromosome Location
Chr.9: 92031134 - 92115474 on GRCh38
UniGene ID
Hs.90458
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001281303.1 NP_001268232.1 4
NM_006415.3 NP_006406.1 4
NM_178324.2 NP_847894.1 4
XM_011518138.2 XP_011516440.1 4
XM_011518139.2 XP_011516441.1 4
XM_017014200.1 XP_016869689.1 4
XM_017014201.1 XP_016869690.1 4