gRNA Design Details

Target DNA Sequence
TAAAGCCGTGCAAGCCTTTG
PAM Sequence
TGG
Target locus
Chr.2: 200412394 - 200412416 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 90.06/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
SPATS2L
Entrez Gene ID
26010
Gene Name
spermatogenesis associated serine rich 2 like
Gene Aliases
DNAPTP6, SGNP
Chromosome Location
Chr.2: 200305881 - 200482263 on GRCh38
UniGene ID
Hs.120323
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001100422.1 NP_001093892.1 4
NM_001100423.1 NP_001093893.1 4
NM_001100424.1 NP_001093894.1 4
NM_001282735.1 NP_001269664.1 4
NM_001282743.1 NP_001269672.1 3
NM_001282744.1 NP_001269673.1 4
NM_015535.2 NP_056350.2 4
XM_005246456.1 XP_005246513.1 5
XM_005246457.1 XP_005246514.1 5
XM_005246458.1 XP_005246515.1 3
XM_005246459.1 XP_005246516.1 4
XM_011510938.1 XP_011509240.1 5
XM_011510939.1 XP_011509241.1 4
XM_017003783.1 XP_016859272.1 5
XM_017003784.1 XP_016859273.1 4
XM_017003785.1 XP_016859274.1 3
XM_017003786.1 XP_016859275.1 5
XM_017003787.1 XP_016859276.1 5