Targeted Isoforms | Translated Protein | Targeted Exon(s) | |
---|---|---|---|
RefSeq | NM_001114093.1 | NP_001107565.1 | 2 |
NM_015578.2 | NP_056393.2 | 2 | |
XM_005258719.4 | XP_005258776.1 | 2 | |
XM_005258720.4 | XP_005258777.1 | 2 | |
XM_005258721.4 | XP_005258778.1 | 2 | |
XM_011526708.2 | XP_011525010.1 | 2 | |
XM_017026575.1 | XP_016882064.1 | 2 | |
XM_017026576.1 | XP_016882065.1 | 2 | |
XM_017026577.1 | XP_016882066.1 | 2 |