gRNA Design Details

Target DNA Sequence
CTTGACGCCGTTGAAGTACT
PAM Sequence
CGG
Target locus
Chr.7: 30681928 - 30681950 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 97.27/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
CRHR2
Entrez Gene ID
1395
Gene Name
corticotropin releasing hormone receptor 2
Gene Aliases
CRF-RB, CRF2, CRFR2, HM-CRF
Chromosome Location
Chr.7: 30651844 - 30700103 on GRCh38
UniGene ID
Hs.546246
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001202475.1 NP_001189404.1 3
NM_001202481.1 NP_001189410.1 4
NM_001202482.1 NP_001189411.1 2
NM_001202483.1 NP_001189412.1 2
NM_001883.4 NP_001874.2 2
XM_011515127.2 XP_011513429.1 2
XM_011515128.2 XP_011513430.1 2
XM_011515129.2 XP_011513431.1 2
XM_017011751.1 XP_016867240.1 2
XM_017011752.1 XP_016867241.1 2