gRNA Design Details
- Target DNA Sequence
- TGCACCACGCTACCATGAAG
- PAM Sequence
- CGG
- Target locus
-
Chr.14: 38255882 - 38255904 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
90.19/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CLEC14A
- Entrez Gene ID
-
161198
- Gene Name
- C-type lectin domain family 14 member A
- Gene Aliases
-
C14orf27, CEG1, EGFR-5
- Chromosome Location
-
Chr.14: 38254000 - 38256370 on GRCh38
- UniGene ID
- Hs.525307
- Species
-
Human