gRNA Design Details
- Target DNA Sequence
- TAGGCTCACTCCCCTTCTTA
- PAM Sequence
- TGG
- Target locus
-
Chr.4: 90308497 - 90308519 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
89.5/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CCSER1
- Entrez Gene ID
-
401145
- Gene Name
- coiled-coil serine rich protein 1
- Gene Aliases
-
FAM190A
- Chromosome Location
-
Chr.4: 90127394 - 91602219 on GRCh38
- UniGene ID
- Hs.654735
- Species
-
Human