gRNA Design Details
- Target DNA Sequence
- GTCTTCGGAGTGCAACTCTG
- PAM Sequence
- AGG
- Target locus
-
Chr.19: 18386317 - 18386339 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
90.14/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GDF15
- Entrez Gene ID
-
9518
- Gene Name
- growth differentiation factor 15
- Gene Aliases
-
GDF-15, MIC-1, MIC1, NAG-1, PDF, PLAB, PTGFB
- Chromosome Location
-
Chr.19: 18386158 - 18389176 on GRCh38
- UniGene ID
- Hs.616962
- Species
-
Human