gRNA Design Details

Target DNA Sequence
GAATATGATGAGGACGACCT
PAM Sequence
GGG
Target locus
Chr.17: 36952948 - 36952970 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 90.17/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
AATF
Entrez Gene ID
26574
Gene Name
apoptosis antagonizing transcription factor
Gene Aliases
BFR2, CHE-1, CHE1, DED
Chromosome Location
Chr.17: 36948875 - 37056871 on GRCh38
UniGene ID
Hs.195740
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_012138.3 NP_036270.1 3
XM_011524611.1 XP_011522913.1 3