gRNA Design Details

Target DNA Sequence
ACCTTTTGTCCGTGTCCAAG
PAM Sequence
AGG
Target locus
Chr.17: 36950311 - 36950333 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 90.63/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
AATF
Entrez Gene ID
26574
Gene Name
apoptosis antagonizing transcription factor
Gene Aliases
BFR2, CHE-1, CHE1, DED
Chromosome Location
Chr.17: 36948875 - 37056871 on GRCh38
UniGene ID
Hs.195740
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_012138.3 NP_036270.1 2
XM_011524611.1 XP_011522913.1 2