gRNA Design Details

Target DNA Sequence
CATCTGCAAGGAATACGTAG
PAM Sequence
AGG
Target locus
Chr.9: 96654737 - 96654759 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 89.55/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
AAED1
Entrez Gene ID
195827
Gene Name
AhpC/TSA antioxidant enzyme domain containing 1
Gene Aliases
C9orf21
Chromosome Location
Chr.9: 96641251 - 96655331 on GRCh38
UniGene ID
Hs.44640
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_153698.1 NP_714542.1 2
XM_005251783.3 XP_005251840.1 2
XM_005251784.4 XP_005251841.1 2