gRNA Design Details
- Target DNA Sequence
- CATCTGCAAGGAATACGTAG
- PAM Sequence
- AGG
- Target locus
-
Chr.9: 96654737 - 96654759 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
89.55/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- AAED1
- Entrez Gene ID
-
195827
- Gene Name
- AhpC/TSA antioxidant enzyme domain containing 1
- Gene Aliases
-
C9orf21
- Chromosome Location
-
Chr.9: 96641251 - 96655331 on GRCh38
- UniGene ID
- Hs.44640
- Species
-
Human