gRNA Design Details

Target DNA Sequence
TACGAAGTAACGGGTGTAGT
PAM Sequence
AGG
Target locus
Chr.16: 2818760 - 2818782 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 95.64/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
PRSS21
Entrez Gene ID
10942
Gene Name
protease, serine 21
Gene Aliases
ESP-1, ESP1, TEST1, TESTISIN
Chromosome Location
Chr.16: 2817163 - 2821722 on GRCh38
UniGene ID
Hs.72026
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001270452.1 NP_001257381.1 4
NM_006799.3 NP_006790.1 4
NM_144956.2 NP_659205.1 4
NM_144957.2 NP_659206.1 4
NR_073012.1 4
XM_017022875.1 XP_016878364.1 4
XM_017022876.1 XP_016878365.1 4
XM_017022877.1 XP_016878366.1 4
XM_017022878.1 XP_016878367.1 4