gRNA Design Details

Target DNA Sequence
GACACCTGAGCAGAAGGAAT
PAM Sequence
GGG
Target locus
Chr.2: 241142874 - 241142896 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 85.94/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
PASK
Entrez Gene ID
23178
Gene Name
PAS domain containing serine/threonine kinase
Gene Aliases
PASKIN, STK37
Chromosome Location
Chr.2: 241106099 - 241150347 on GRCh38
UniGene ID
Hs.397891
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001252119.1 NP_001239048.1 2
NM_001252120.1 NP_001239049.1 3
NM_001252122.1 NP_001239051.1 2
NM_001252124.1 NP_001239053.1 2
NM_015148.3 NP_055963.2 2
XM_011510826.1 XP_011509128.1 2
XM_011510828.1 XP_011509130.1 3
XM_011510829.1 XP_011509131.1 2
XM_011510830.1 XP_011509132.1 2
XM_011510831.2 XP_011509133.1 2
XM_011510832.2 XP_011509134.1 2
XM_017003634.1 XP_016859123.1 2
XM_017003635.1 XP_016859124.1 3
XM_017003636.1 XP_016859125.1 2