gRNA Design Details

Target DNA Sequence
CTCCACCAGTAGGACATTCC
PAM Sequence
CGG
Target locus
Chr.3: 129171688 - 129171710 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 88.57/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
CNBP
Entrez Gene ID
7555
Gene Name
CCHC-type zinc finger nucleic acid binding protein
Gene Aliases
CNBP1, DM2, PROMM, RNF163, ZCCHC22, ZNF9
Chromosome Location
Chr.3: 129167815 - 129183967 on GRCh38
UniGene ID
Hs.518249
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001127192.1 NP_001120664.1 2
NM_001127193.1 NP_001120665.1 2
NM_001127194.1 NP_001120666.1 2
NM_001127195.1 NP_001120667.1 2
NM_001127196.1 NP_001120668.1 2
NM_003418.4 NP_003409.1 2