gRNA Design Details
- Target DNA Sequence
- ACATTGAGGCACTGAACCTG
- PAM Sequence
- GGG
- Target locus
-
Chr.8: 8892846 - 8892868 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
86.47/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- MFHAS1
- Entrez Gene ID
-
9258
- Gene Name
- malignant fibrous histiocytoma amplified sequence 1
- Gene Aliases
-
LRRC65, MASL1
- Chromosome Location
-
Chr.8: 8783354 - 8893621 on GRCh38
- UniGene ID
- Hs.379414
- Species
-
Human