gRNA Design Details
- Target DNA Sequence
- ATAGCCAGATGAAGGGAGCG
- PAM Sequence
- AGG
- Target locus
-
Chr.19: 12843555 - 12843577 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
86.54/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- MAST1
- Entrez Gene ID
-
22983
- Gene Name
- microtubule associated serine/threonine kinase 1
- Gene Aliases
-
SAST, SAST170
- Chromosome Location
-
Chr.19: 12833931 - 12874953 on GRCh38
- UniGene ID
- Hs.227489
- Species
-
Human