gRNA Design Details
- Target DNA Sequence
- GTAAATGTCGGTGACACTCT
- PAM Sequence
- TGG
- Target locus
-
Chr.3: 39266303 - 39266325 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
92.38/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CX3CR1
- Entrez Gene ID
-
1524
- Gene Name
- C-X3-C motif chemokine receptor 1
- Gene Aliases
-
CCRL1, CMKBRL1, CMKDR1, GPR13, GPRV28, V28
- Chromosome Location
-
Chr.3: 39263494 - 39281735 on GRCh38
- UniGene ID
- Hs.78913
- Species
-
Human