Targeted Isoforms | Translated Protein | Targeted Exon(s) | |
---|---|---|---|
RefSeq | NM_032291.3 | NP_115667.2 | 4 |
XM_006710967.2 | XP_006711030.1 | 4 | |
XM_011542291.1 | XP_011540593.1 | 4 | |
XM_011542292.1 | XP_011540594.1 | 4 | |
XM_017002505.1 | XP_016857994.1 | 4 | |
XM_017002506.1 | XP_016857995.1 | 5 | |
XM_017002507.1 | XP_016857996.1 | 4 | |
XM_017002508.1 | XP_016857997.1 | 5 | |
XM_017002509.1 | XP_016857998.1 | 4 | |
XM_017002510.1 | XP_016857999.1 | 5 | |
XM_017002511.1 | XP_016858000.1 | 5 | |
XM_017002512.1 | XP_016858001.1 | 5 | |
XM_017002513.1 | XP_016858002.1 | 4 | |
XM_017002517.1 | XP_016858006.1 | 5 | |
XM_017002524.1 | XP_016858013.1 | 5 | |
XM_017002525.1 | XP_016858014.1 | 5 | |
XM_017002526.1 | XP_016858015.1 | 4 | |
XM_017002527.1 | XP_016858016.1 | 5 | |
XM_017002530.1 | XP_016858019.1 | 4 | |
XM_017002533.1 | XP_016858022.1 | 5 |