gRNA Design Details
- Target DNA Sequence
- GCAGCTGATATTCTTCGACT
- PAM Sequence
- GGG
- Target locus
-
Chr.6: 47025980 - 47026002 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
91.68/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- ADGRF1
- Entrez Gene ID
-
266977
- Gene Name
- adhesion G protein-coupled receptor F1
- Gene Aliases
-
GPR110, KPG_012, PGR19, hGPCR36
- Chromosome Location
-
Chr.6: 46997703 - 47042363 on GRCh38
- UniGene ID
- Hs.733762
- Species
-
Human