gRNA Design Details

Target DNA Sequence
CGTAGCGCTGGATCCATTCA
PAM Sequence
AGG
Target locus
Chr.14: 24206613 - 24206635 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 92.32/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
TSSK4
Entrez Gene ID
283629
Gene Name
testis specific serine kinase 4
Gene Aliases
C14orf20, STK22E, TSK-4, TSK4, TSSK5
Chromosome Location
Chr.14: 24205524 - 24208362 on GRCh38
UniGene ID
Hs.314432
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001184739.1 NP_001171668.1 2
NM_001308067.1 NP_001294996.1 2
NM_174944.3 NP_777604.2 2
XM_011536663.2 XP_011534965.1 2
XM_017021226.1 XP_016876715.1 2