gRNA Design Details
- Target DNA Sequence
- AGCCTCATATACCGACCCAT
- PAM Sequence
- AGG
- Target locus
-
Chr.14: 24206027 - 24206049 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
92.49/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TSSK4
- Entrez Gene ID
-
283629
- Gene Name
- testis specific serine kinase 4
- Gene Aliases
-
C14orf20, STK22E, TSK-4, TSK4, TSSK5
- Chromosome Location
-
Chr.14: 24205524 - 24208362 on GRCh38
- UniGene ID
- Hs.314432
- Species
-
Human