gRNA Design Details
- Target DNA Sequence
- TGGAAAACTGTTCTGTCCCC
- PAM Sequence
- AGG
- Target locus
-
Chr.5: 148070358 - 148070380 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
83.86/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SPINK5
- Entrez Gene ID
-
11005
- Gene Name
- serine peptidase inhibitor, Kazal type 5
- Gene Aliases
-
LEKTI, LETKI, NETS, NS, VAKTI
- Chromosome Location
-
Chr.5: 148063972 - 148137362 on GRCh38
- UniGene ID
- Hs.331555
- Species
-
Human