gRNA Design Details

Target DNA Sequence
CCTGCTCTACATCAACCCTG
PAM Sequence
AGG
Target locus
Chr.2: 49017520 - 49017542 on GRCh38
Strand
Reverse
Application
Gene Knockout
Score: 86.86/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
FSHR
Entrez Gene ID
2492
Gene Name
follicle stimulating hormone receptor
Gene Aliases
FSHR1, FSHRO, LGR1, ODG1
Chromosome Location
Chr.2: 48953161 - 49154527 on GRCh38
UniGene ID
Hs.1428
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_000145.3 NP_000136.2 4
NM_181446.2 NP_852111.2 4
XM_011532733.2 XP_011531035.1 4
XM_011532734.2 XP_011531036.1 4
XM_011532740.1 XP_011531042.1 4