gRNA Design Details

Target DNA Sequence
TCAGTTCCCATCTAGCAAGG
PAM Sequence
TGG
Target locus
Chr.16: 31474165 - 31474187 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 86.54/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
TGFB1I1
Entrez Gene ID
7041
Gene Name
transforming growth factor beta 1 induced transcript 1
Gene Aliases
ARA55, HIC-5, HIC5, TSC-5
Chromosome Location
Chr.16: 31472155 - 31477960 on GRCh38
UniGene ID
Hs.513530
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001042454.2 NP_001035919.1 5
NM_001164719.1 NP_001158191.1 5
NM_015927.4 NP_057011.2 5