gRNA Design Details
- Target DNA Sequence
- TCCTCTTCCAGCGGTGTCTT
- PAM Sequence
- GGG
- Target locus
-
Chr.16: 31473890 - 31473912 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
86.92/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- TGFB1I1
- Entrez Gene ID
-
7041
- Gene Name
- transforming growth factor beta 1 induced transcript 1
- Gene Aliases
-
ARA55, HIC-5, HIC5, TSC-5
- Chromosome Location
-
Chr.16: 31472155 - 31477960 on GRCh38
- UniGene ID
- Hs.513530
- Species
-
Human