Targeted Isoforms | Translated Protein | Targeted Exon(s) | |
---|---|---|---|
RefSeq | NM_001198777.1 | NP_001185706.1 | 2 |
NM_001198778.1 | NP_001185707.1 | 2 | |
NM_001198779.1 | NP_001185708.1 | 2 | |
NM_001324375.1 | NP_001311304.1 | 2 | |
NM_001324376.1 | NP_001311305.1 | 2 | |
NM_003591.3 | NP_003582.2 | 2 | |
XM_011519743.1 | XP_011518045.1 | 4 | |
XM_011519744.1 | XP_011518046.1 | 3 | |
XM_011519745.1 | XP_011518047.1 | 3 | |
XM_011519747.1 | XP_011518049.1 | 3 |