gRNA Design Details
- Target DNA Sequence
- ATCACATACCTACCAGTTTG
- PAM Sequence
- TGG
- Target locus
-
Chr.5: 148314151 - 148314173 on GRCh38
- Strand
- Forward
- Application
- Gene Knockout
Score:
87.56/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- SPINK7
- Entrez Gene ID
-
84651
- Gene Name
- serine peptidase inhibitor, Kazal type 7 (putative)
- Gene Aliases
-
ECG2, ECRG2
- Chromosome Location
-
Chr.5: 148312427 - 148315919 on GRCh38
- UniGene ID
- Hs.244569
- Species
-
Human