gRNA Design Details
- Target DNA Sequence
- TGAGCAACCAAACGCGGAGG
- PAM Sequence
- TGG
- Target locus
-
Chr.9: 108826622 - 108826644 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
88.63/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Celsr3
- Entrez Gene ID
-
107934
- Gene Name
- cadherin, EGF LAG seven-pass G-type receptor 3 (flamingo homolog, Drosophila)
- Gene Aliases
-
Adgrc3, Fmi1, flamingo, mKIAA0812
- Chromosome Location
-
Chr.9: 108826045 - 108852969 on GRCm38
- UniGene ID
- Mm.39945
- Species
-
Mouse