gRNA Design Details
- Target DNA Sequence
- GTCTTCATGAGGCTCCGAGT
- PAM Sequence
- AGG
- Target locus
-
Chr.9: 108826566 - 108826588 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
86.17/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Celsr3
- Entrez Gene ID
-
107934
- Gene Name
- cadherin, EGF LAG seven-pass G-type receptor 3 (flamingo homolog, Drosophila)
- Gene Aliases
-
Adgrc3, Fmi1, flamingo, mKIAA0812
- Chromosome Location
-
Chr.9: 108826045 - 108852969 on GRCm38
- UniGene ID
- Mm.39945
- Species
-
Mouse