gRNA Design Details
- Target DNA Sequence
- GCCACTGATAGGAACCCTAG
- PAM Sequence
- AGG
- Target locus
-
Chr.4: 80266930 - 80266952 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
90.02/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- FGF5
- Entrez Gene ID
-
2250
- Gene Name
- fibroblast growth factor 5
- Gene Aliases
-
HBGF-5, Smag-82, TCMGLY
- Chromosome Location
-
Chr.4: 80266588 - 80291017 on GRCh38
- UniGene ID
- Hs.37055
- Species
-
Human