gRNA Design Details

Target DNA Sequence
GGATAGAGGGTAATGAAAGC
PAM Sequence
TGG
Target locus
Chr.4: 54261114 - 54261136 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 87.67/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
PDGFRA
Entrez Gene ID
5156
Gene Name
platelet derived growth factor receptor alpha
Gene Aliases
CD140A, GAS9, PDGFR-2, PDGFR2, RHEPDGFRA
Chromosome Location
Chr.4: 54229097 - 54298245 on GRCh38
UniGene ID
Hs.74615
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_006206.4 NP_006197.1 3
XM_005265743.1 XP_005265800.1 3
XM_006714039.3 XP_006714102.1 4
XM_006714041.3 XP_006714104.1 4
XM_011534385.2 XP_011532687.1 3
XM_017008280.1 XP_016863769.1 3
XM_017008281.1 XP_016863770.1 3
XM_017008282.1 XP_016863771.1 3