gRNA Design Details

Target DNA Sequence
ATCTACGTGCACAACAGCCC
PAM Sequence
TGG
Target locus
Chr.1: 43305141 - 43305163 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 88.99/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
TIE1
Entrez Gene ID
7075
Gene Name
tyrosine kinase with immunoglobulin like and EGF like domains 1
Gene Aliases
JTK14, TIE
Chromosome Location
Chr.1: 43300895 - 43323110 on GRCh38
UniGene ID
Hs.78824
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001253357.1 NP_001240286.1 2
NM_005424.4 NP_005415.1 2
XM_005271163.1 XP_005271220.1 2
XM_006710869.1 XP_006710932.1 2
XM_017002207.1 XP_016857696.1 2