gRNA Design Details
- Target DNA Sequence
- AAAGGCGCAAATGCCGGGTC
- PAM Sequence
- AGG
- Target locus
-
Chr.22: 17008050 - 17008072 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
90.36/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- GAB4
- Entrez Gene ID
-
128954
- Gene Name
- GRB2 associated binding protein family member 4
- Gene Aliases
-
-
- Chromosome Location
-
Chr.22: 16961936 - 17008281 on GRCh38
- UniGene ID
- Hs.434113
- Species
-
Human