gRNA Design Details

Target DNA Sequence
GTTCAGGTTGATGGTGCGCA
PAM Sequence
GGG
Target locus
Chr.22: 16992060 - 16992082 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 88.31/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
GAB4
Entrez Gene ID
128954
Gene Name
GRB2 associated binding protein family member 4
Gene Aliases
-
Chromosome Location
Chr.22: 16961936 - 17008281 on GRCh38
UniGene ID
Hs.434113
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001037814.1 NP_001032903.1 2
XM_011546114.2 XP_011544416.1 2
XM_011546115.1 XP_011544417.1 2
XM_011546116.2 XP_011544418.1 2
XM_017028575.1 XP_016884064.1 2