gRNA Design Details
- Target DNA Sequence
- TGTACTCAACGCCTCCACCT
- PAM Sequence
- TGG
- Target locus
-
Chr.19: 18599673 - 18599695 on GRCh38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
86.42/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- CRLF1
- Entrez Gene ID
-
9244
- Gene Name
- cytokine receptor like factor 1
- Gene Aliases
-
CISS, CISS1, CLF, CLF-1, NR6, zcytor5
- Chromosome Location
-
Chr.19: 18593225 - 18606850 on GRCh38
- UniGene ID
- Hs.114948
- Species
-
Human