gRNA Design Details

Target DNA Sequence
CATCTTCCATGTATGAATGG
PAM Sequence
TGG
Target locus
Chr.2: 33186864 - 33186886 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 87.62/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
LTBP1
Entrez Gene ID
4052
Gene Name
latent transforming growth factor beta binding protein 1
Gene Aliases
-
Chromosome Location
Chr.2: 32947129 - 33399509 on GRCh38
UniGene ID
Hs.619315
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_000627.3 NP_000618.3 2
NM_001166264.1 NP_001159736.1 2
NM_001166265.1 NP_001159737.1 2
NM_001166266.1 NP_001159738.1 2
NM_206943.2 NP_996826.2 6
XM_005264317.3 XP_005264374.1 6
XM_005264318.3 XP_005264375.1 6
XM_011532853.2 XP_011531155.1 6
XM_011532855.2 XP_011531157.1 6
XM_011532856.2 XP_011531158.1 6
XM_011532857.2 XP_011531159.1 6
XM_011532858.2 XP_011531160.1 6
XM_011532859.2 XP_011531161.1 6
XM_011532860.2 XP_011531162.1 5
XM_011532861.2 XP_011531163.1 6
XM_011532862.2 XP_011531164.1 6
XM_017004108.1 XP_016859597.1 6
XM_017004109.1 XP_016859598.1 6
XM_017004110.1 XP_016859599.1 6