gRNA Design Details

Target DNA Sequence
TGGCTTCTGGTGTTGGCAGT
PAM Sequence
GGG
Target locus
Chr.9: 136668313 - 136668335 on GRCh38
Strand
Forward
Application
Gene Knock-Out
Score: 80.9/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
EGFL7
Entrez Gene ID
51162
Gene Name
EGF like domain multiple 7
Gene Aliases
NEU1, VE-STATIN, ZNEU1
Chromosome Location
Chr.9: 136654753 - 136672678 on GRCh38
UniGene ID
Hs.91481
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_016215.4 NP_057299.1 4
NM_201446.2 NP_958854.1 2
NR_045110.1 3
NR_045111.1 3
NR_046367.1 3
XM_006717141.3 XP_006717204.1 2
XM_011518764.1 XP_011517066.1 4
XM_011518765.1 XP_011517067.1 4
XM_011518766.1 XP_011517068.1 5
XM_017014797.1 XP_016870286.1 4